MAT390510318 Matrix Scientific

Ammonium 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonate

  • MDL number MFCD12922538
  • Cas Number : 29081-56-9
  • Molecular Weight : 517.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/099009.html

Safety Information

MSDS http://www.matrixscientific.com/099009.html
pictograms Marine Pollutant/Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart