MAT390510379 Matrix Scientific

Cholesteryl chloroformate

  • MDL number MFCD00003633
  • Cas Number : 7144-08-3
  • Molecular Weight : 449.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-118
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/099070.html

Safety Information

MSDS http://www.matrixscientific.com/099070.html
pictograms Corrosive
Availability details for
Quantity
  • Availability
  • Add to Cart