MAT390510458 Matrix Scientific

3-Chloropropyl carbonochloridate

  • MDL number MFCD02093717
  • Cas Number : 628-11-5
  • Molecular Weight : 157

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.302
Flash Point 96.1
Country of Origin USA
Boiling point 184-189
Product Link http://www.matrixscientific.com/099149.html

Safety Information

MSDS http://www.matrixscientific.com/099149.html
pictograms Toxic/Corrosive/Lachrymator
Availability details for
Quantity
  • Availability
  • Add to Cart