MAT390510464 Matrix Scientific

Dinonyl phthalate

  • MDL number MFCD00036237
  • Cas Number : 84-76-4
  • Molecular Weight : 418.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 0.97
Flash Point 216
Country of Origin USA
Product Link http://www.matrixscientific.com/099155.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/099155.html
Availability details for
Quantity
  • Availability
  • Add to Cart