MAT390510591 Matrix Scientific

Sulbutiamine

  • MDL number MFCD01726427
  • Cas Number : 3286-46-2
  • Molecular Weight : 702.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerate
Country of Origin USA
Boiling point 892.5 �t 760 mmHg
Product Link http://www.matrixscientific.com/099282.html

Safety Information

MSDS http://www.matrixscientific.com/099282.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart