MAT390510731 Matrix Scientific

Prednisolone phosphate sodium

  • MDL number MFCD00083682
  • Cas Number : 125-02-0
  • Molecular Weight : 484.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/099422.html

Safety Information

MSDS http://www.matrixscientific.com/099422.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart