MAT390510865 Matrix Scientific

Rose Bengal sodium salt

  • MDL number MFCD00151169
  • Cas Number : 632-69-9
  • Molecular Weight : 1017.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 91
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/099556.html

Safety Information

MSDS http://www.matrixscientific.com/099556.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart