MAT390511133 Matrix Scientific

Sodium 4-nitrophenolate

  • MDL number MFCD00044673
  • Cas Number : 824-78-2
  • Molecular Weight : 161.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/099824.html

Safety Information

MSDS http://www.matrixscientific.com/099824.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart