MAT390511170 Matrix Scientific

Sucrose stearate

  • MDL number MFCD00152822
  • Cas Number : 25168-73-4
  • Molecular Weight : 608.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67-71
Purity 95
Density 1.246g/cc
Flash Point 235
Country of Origin USA
Product Link http://www.matrixscientific.com/099861.html

Safety Information

MSDS http://www.matrixscientific.com/099861.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart