MAT390511178 Matrix Scientific

(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)-pentanamide dihydrochloride

  • MDL number MFCD00058249
  • Cas Number : 40127-11-5
  • Molecular Weight : 367.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/099869.html

Safety Information

MSDS http://www.matrixscientific.com/099869.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart