MAT390511186 Matrix Scientific

Bis(4-octylphenyl)amine

  • MDL number MFCD00048942
  • Cas Number : 101-67-7
  • Molecular Weight : 393.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-97
Purity 90
Density 0.939
Flash Point 270
Storage temp. Ambient
Country of Origin USA
Boiling point 509
Product Link http://www.matrixscientific.com/099877.html

Safety Information

MSDS http://www.matrixscientific.com/099877.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart