MAT390511201 Matrix Scientific

(S)-2-Aminosuccinic acid, sodium salt

  • MDL number MFCD00082327
  • Cas Number : 3792-50-5
  • Molecular Weight : 177.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/099892.html

Safety Information

MSDS http://www.matrixscientific.com/099892.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart