MAT390511240 Matrix Scientific

5-O-((1,1-Dimethylethyl)dimethylsilyl)-2,3-O-(1-methylethylidene)-D-ribose

  • MDL number MFCD01075716
  • Cas Number : 68703-51-5
  • Molecular Weight : 304.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-51
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/099931.html

Safety Information

MSDS http://www.matrixscientific.com/099931.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart