MAT390511264 Matrix Scientific

Tetraammineplatinum (II) chloride hydrate

  • MDL number MFCD00149947
  • Cas Number : 13933-33-0
  • Molecular Weight : 352.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/099955.html

Safety Information

MSDS http://www.matrixscientific.com/099955.html
pictograms Irritant/Hygroscopic
Availability details for
Quantity
  • Availability
  • Add to Cart