MAT390511394 Matrix Scientific

(6-Carbamoylpyridin-3-yl)boronic acid

  • MDL number MFCD12964543
  • Cas Number : 1164100-82-6
  • Molecular Weight : 165.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/100087.html

Safety Information

MSDS http://www.matrixscientific.com/100087.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart