MAT390511595 Matrix Scientific

Complanatoside A

  • MDL number MFCD20527287
  • Cas Number : 116183-66-5
  • Molecular Weight : 624.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/100275.html

Safety Information

MSDS http://www.matrixscientific.com/100275.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart