MAT390511645 Matrix Scientific

(5-Bromo-3-fluoro-2-methoxyphenyl)methanol

  • MDL number MFCD23701710
  • Cas Number : 470668-69-0
  • Molecular Weight : 235.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/100325.html

Safety Information

MSDS http://www.matrixscientific.com/100325.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart