MAT390512312 Matrix Scientific

3-Thien-2-ylbenzaldehyde

  • MDL number MFCD04038019
  • Cas Number : 103668-99-1
  • Molecular Weight : 188.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/100992.html

Safety Information

MSDS http://www.matrixscientific.com/100992.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart