MAT390512512 Matrix Scientific

Ethyl 1,3-benzothiazol-2-ylacetate

  • MDL number MFCD00546397
  • Cas Number : 29182-42-1
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/101192.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart