MAT398830196 Matrix Scientific

4-(Pentafluoroethyl)benzylamine

  • MDL number MFCD22416533
  • Cas Number : 128273-62-1
  • Molecular Weight : 225.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/101343.html

Safety Information

MSDS http://www.matrixscientific.com/101343.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart