MAT405191913 Matrix Scientific

2-((S)-2-Benzyl-3-((3R,4R)-4-(3-hydroxyphenyl)-3,4-dimethylpiperidin-1-yl)propanamido)acetic acid

  • MDL number MFCD09838268
  • Cas Number : 156053-89-3
  • Molecular Weight : 461.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/100260.html

Safety Information

MSDS http://www.matrixscientific.com/100260.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart