MAT405191988 Matrix Scientific

4-Iodo-3-(4-methoxyphenyl)-3H-1,2,3-oxadiazol-1-ium-5-olate

  • MDL number MFCD25948963
  • Cas Number : 201282-63-5
  • Molecular Weight : 318.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-71
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/101493.html

Safety Information

MSDS http://www.matrixscientific.com/101493.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart