MAT405192110 Matrix Scientific

3,5-Dichloro-4-formylbenzonitrile

  • MDL number MFCD16251144
  • Cas Number : 157870-18-3
  • Molecular Weight : 200.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/101626.html

Safety Information

MSDS http://www.matrixscientific.com/101626.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart