MAT410320658 Matrix Scientific

1-(1-Fluorocyclopropyl)ethanone

  • MDL number MFCD18205953
  • Cas Number : 63141-10-6
  • Molecular Weight : 102.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerate
Country of Origin USA
Product Link http://www.matrixscientific.com/102325.html

Safety Information

MSDS http://www.matrixscientific.com/102325.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart