MAT410320691 Matrix Scientific

2-Amino-2-methylpropionitrile hydrochloride

  • MDL number MFCD08456816
  • Cas Number : 50846-36-1
  • Molecular Weight : 120.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >275
Purity 90
Country of Origin USA
Product Link http://www.matrixscientific.com/008325.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008325.html
Availability details for
Quantity
  • Availability
  • Add to Cart