MAT411067180 Matrix Scientific

Methyl 2-amino-2-(4-hydroxyphenyl)acetate

  • MDL number MFCD08063907
  • Cas Number : 43189-12-4
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-199
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/102397.html

Safety Information

MSDS http://www.matrixscientific.com/102397.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart