MAT413148326 Matrix Scientific

3-(Tetrahydro-2-furanylmethoxy)piperidine hydrochloride

  • MDL number MFCD27579019
  • Cas Number : 933736-25-5
  • Molecular Weight : 221.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023166.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023166.html
Availability details for
Quantity
  • Availability
  • Add to Cart