MAT424016460 Matrix Scientific

1,3-Di-tert-butyl 2-methylpropanedioate

  • MDL number MFCD06797699
  • Cas Number : 34812-95-8
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/102760.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/102760.html
Availability details for
Quantity
  • Availability
  • Add to Cart