MAT424016555 Matrix Scientific

6-Chloro-2-iodo-9-(2',3',5'-tri-O-acetyl-beta-D-ribofuranosyl)purine

  • MDL number MFCD03701153
  • Cas Number : 5987-76-8
  • Molecular Weight : 538.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://www.matrixscientific.com/102872.html

Safety Information

MSDS http://www.matrixscientific.com/102872.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart