MAT424017030 Matrix Scientific

2-Ethyl-4'-methoxybenzophenone

  • MDL number MFCD01311579
  • Cas Number : 341022-06-8
  • Molecular Weight : 240.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-35
Country of Origin USA
Product Link http://www.matrixscientific.com/103354.html

Safety Information

msds http://www.matrixscientific.com/103354.html
Availability details for
Quantity
  • Availability
  • Add to Cart