MAT424017169 Matrix Scientific

4-Bromo-4'-tert-butylbenzophenone

  • MDL number MFCD06201443
  • Cas Number : 91404-26-1
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Country of Origin US
Product Link http://www.matrixscientific.com/103493.html

Safety Information

MSDS http://www.matrixscientific.com/103493.html
Availability details for
Quantity
  • Availability
  • Add to Cart