MAT424017174 Matrix Scientific

2-Bromo-3',4'-dimethoxybenzophenone

  • MDL number MFCD02127302
  • Cas Number : 23346-79-4
  • Molecular Weight : 321.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-157
Country of Origin USA
Product Link http://www.matrixscientific.com/103498.html

Safety Information

msds http://www.matrixscientific.com/103498.html
Availability details for
Quantity
  • Availability
  • Add to Cart