MAT424807653 Matrix Scientific

3-[(2-Naphthyloxy)methyl]piperidine hydrochloride

  • MDL number MFCD28023812
  • Cas Number : 1706435-72-4
  • Molecular Weight : 277.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023031.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023031.html
Availability details for
Quantity
  • Availability
  • Add to Cart