MAT424807666 Matrix Scientific

3-Methyl-1H-pyrazol-5-amine dihydrochloride

  • MDL number MFCD17168390
  • Cas Number : 31230-17-8
  • Molecular Weight : 170.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039731.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039731.html
Availability details for
Quantity
  • Availability
  • Add to Cart