MAT424812401 Matrix Scientific

4'-Chloro-3'-fluoro-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843274
  • Cas Number : 898781-42-5
  • Molecular Weight : 308.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/110092.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/110092.html
Availability details for
Quantity
  • Availability
  • Add to Cart