MAT424813646 Matrix Scientific

Ethyl 8-(3-chlorophenyl)-8-oxooctanoate

  • MDL number MFCD01320240
  • Cas Number : 898752-20-0
  • Molecular Weight : 296.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/111337.html

Safety Information

MSDS http://www.matrixscientific.com/111337.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart