MAT428344011 Matrix Scientific

Boc-3-methoxy-L-tyrosine(dicyclohexylammonium) salt

  • MDL number MFCD28099699
  • Cas Number : 53267-94-0
  • Molecular Weight : 492.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/041867.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041867.html
Availability details for
Quantity
  • Availability
  • Add to Cart