MAT428344041 Matrix Scientific

Fmoc-(R)-3-amino-4-(2-thienyl)-butyric acid

  • MDL number MFCD01860985
  • Molecular Weight : 407.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/042305.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042305.html
Availability details for
Quantity
  • Availability
  • Add to Cart