MAT428344042 Matrix Scientific

Fmoc-(S)-3-amino-4-(2-thienyl)-butyric acid

  • MDL number MFCD01861082
  • Cas Number : 270262-98-1
  • Molecular Weight : 407.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/042342.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042342.html
Availability details for
Quantity
  • Availability
  • Add to Cart