MAT428344061 Matrix Scientific

(S)-2-Acetamido-5-guanidinopentanoic acid dihydrate

  • MDL number MFCD00150285
  • Cas Number : 155-84-0
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/093553.html

Safety Information

MSDS http://www.matrixscientific.com/093553.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart