MAT428344091 Matrix Scientific

3-(1,1-Difluoroethyl)aniline

  • MDL number MFCD20483887
  • Cas Number : 40788-05-4
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/113507.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/113507.html
Availability details for
Quantity
  • Availability
  • Add to Cart