MAT428344134 Matrix Scientific

N-(2-Methoxyethyl)ethylamine

  • MDL number MFCD00144876
  • Cas Number : 34322-82-2
  • Molecular Weight : 103.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/113550.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/113550.html
Availability details for
Quantity
  • Availability
  • Add to Cart