MAT428344325 Matrix Scientific

1,1'-(4-Chlorobutylidene)bis(4-fluorobenzene)

  • MDL number MFCD00044723
  • Cas Number : 3312-04-7
  • Molecular Weight : 280.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.2039/20
Flash Point 187
Country of Origin USA
Boiling point 166-168�Torr
Product Link http://www.matrixscientific.com/113746.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart