MAT428344719 Matrix Scientific

3-(3-Bromophenyl)-N-ethylacrylamide

  • MDL number MFCD00075351
  • Cas Number : 58473-74-8
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-91
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/114148.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart