MAT428344888 Matrix Scientific

4-(Trifluoromethyl)phenylacetyl chloride

  • MDL number MFCD08458136
  • Cas Number : 74426-51-0
  • Molecular Weight : 222.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerate/argon
Country of Origin USA
Boiling point 94-97°/10mmHg
Product Link http://www.matrixscientific.com/114323.html

Safety Information

MSDS http://www.matrixscientific.com/114323.html
pictograms Corrosive/Stored under Argon/Refrigerate/Lachrymator
Availability details for
Quantity
  • Availability
  • Add to Cart