MAT428345113 Matrix Scientific

4-(Pentafluorosulfanyl)phenyl isocyanate

  • MDL number MFCD28099770
  • Cas Number : 159689-43-7
  • Molecular Weight : 245.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97

Safety Information

msds http://www.matrixscientific.com/114803.html
Availability details for
Quantity
  • Availability
  • Add to Cart