MAT428345124 Matrix Scientific

2-(4-Chloro-2-iodophenoxy)-3-nitropyridine

  • MDL number MFCD28099779
  • Cas Number : 1706438-57-4
  • Molecular Weight : 376.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/114945.html

Safety Information

MSDS http://www.matrixscientific.com/114945.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart