MAT428345158 Matrix Scientific

2-Chloro-4-methoxynicotinic acid

  • MDL number MFCD18378504
  • Cas Number : 394729-98-7
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97

Safety Information

msds http://www.matrixscientific.com/114527.html
Availability details for
Quantity
  • Availability
  • Add to Cart