MAT428345349 Matrix Scientific

Ethyl 5-bromo-1,3,4-thiadiazole-2-carboxylate

  • MDL number MFCD12165919
  • Cas Number : 1030613-07-0
  • Molecular Weight : 237.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerate/dark
Country of Origin USA
Product Link http://www.matrixscientific.com/114718.html

Safety Information

hcodes Irritant-Harmful/Refrigerate/Light Sensitive
Availability details for
Quantity
  • Availability
  • Add to Cart