MAT428345443 Matrix Scientific

4-(Trifluoromethyl)pyrimidine

  • MDL number MFCD13186003
  • Cas Number : 136547-16-5
  • Molecular Weight : 148.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/114812.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/114812.html
Availability details for
Quantity
  • Availability
  • Add to Cart